Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1279-01

rProtein A Sepharose Fast Flow

Cytiva 17-1279-01, pack of 5 mL

Synonym(s):

Fast Flow Sepharose, Protein A Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

recombinant protein A (E. coli)

packaging

pack of 5 mL

manufacturer/tradename

Cytiva 17-1279-01

storage condition

(20% Ehtanol)

matrix

4% cross-linked agarose

particle size

60-165 μm

average diameter

90 μm (d50v)

cleaning in place

2-11

working range

3-10

capacity

~50 mg binding capacity (human lgG/ml medium)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuxiao Wang et al.
Journal of orthopaedic translation, 35, 72-80 (2022-10-04)
Bone fracture healing is a postnatal regenerative process in which fibrocartilaginous callus formation and bony callus formation are important. Bony callus formation requires osteoblastic differentiation of MSCs. The formation of callus was assessed by μCT, Safranin-O, H&E and Masson trichrome
Jeannine A Frei et al.
eNeuro, 8(4) (2021-06-18)
Autism spectrum disorder (ASD) is a neurologic condition characterized by alterations in social interaction and communication, and restricted and/or repetitive behaviors. The classical Type II cadherins cadherin-8 (Cdh8, CDH8) and cadherin-11 (Cdh11, CDH11) have been implicated as autism risk gene

Articles

This page shows various purification options for Protein A Sepharose chromatography media and describes typical binding and elution conditions for Protein A Sepharose chromatography media.

This page describes immunoprecipitation (immunoaffinity or pull-down techniques).

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

Protocols

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service