Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0468-01

Agarose IEF

Cytiva 17-0468-01, pack of 10 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pack of 10 g

manufacturer/tradename

Cytiva 17-0468-01

storage condition

dry at room temperature

color

White

General description

DNA/RNA techniques
Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications. If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used. For preparative work, an agarose with low gelling temperature should be used. Agarose Prep has both low melting and gelling points

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service