Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0402-01

HiTrap® Protein A High Performance

Cytiva 17-0402-01, pack of 5 × 1 mL

Synonym(s):

Protein A Affinity Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

native protein A

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-0402-01

storage condition

(20% Ethanol)

parameter

flow rate
<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-10

working range

3-9

storage temp.

2-8°C

Related Categories

General description

HiTrap® Protein A HP is a prepacked Protein A Sepharose High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites, serum, and cell culture supernatants.

HiTrap® Protein A HP is a prepacked ready to use, column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column, together with the medium provide fast, simple and easy separations in a convenient format.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • High binding capacitiy of 20 mg IgG/mL medium and fast kinetics.
  • 34 μm bead size for high-performance, high-resolution purifications.
  • Designed for use with syringe, pump, or chromatography system such as AKTA design or FPLC System.
  • Stable over pH 3 to 9 for use, and 2 to 10 for cleaning.
  • Robust, easy-to-use method.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

FPLC is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to purify IgM antibodies by affinity chromatography using HiTrap™ IgM Purification HP.

This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.

Protocols

General instructions for affinity purification using HiTrap columns from Cytiva.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service