Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE10600012

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.45 μm, roll W × L 150 mm × 4 m, pkg of 1 ea

Synonym(s):

Nitrocellulose Blotting Membrane, Protran Nitrocellulose Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600012

roll W × L

150 mm × 4 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Nitrocellulose (NC) membrane for use in Western blot protocols, offering low background and high signal-to-noise ratios. Amersham Protran 0.45 NC is a nitrocellulose membrane with a 0.45 µm pore size for use in Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background noise, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting detection reagents. Protran NC membranes are available in popular precut sizes for a wide range of laboratory applications. This product was formerly called Protran BA85.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Offers low background, high signal-to-noise ratios, and high binding capacity
  • Compatible with various labeling and detection systems
  • Use for proteins with a wide range of molecular weights
  • A perfect tool for the purification of antibody and analysis workflow
  • Proprietary nitrocellulose formulation offers a long shelf life of bound proteins

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service