Key Documents
GBL805025
Grace Bio-Labs PATH® protein microarray slides, no barcode
Synonym(s):
non-porous nitrocellulose film slide
About This Item
UNSPSC Code:
41100000
NACRES:
NB.15
material
nitrocellulose (on glass slide)
packaging
pack of 20 ea
manufacturer/tradename
Grace Bio-Labs 805025
external size
25 mm × 75 mm
General description
The Grace Bio-Labs PATH protein microarray slides are a non-porous nitrocellulose film slides.
Benefits:
  • Low and consistant fluorescent backgrounds
  • Higher protein binding
  • Superior signal-to-noise
Legal Information
PATH is a registered trademark of Courtagen Life Sciences, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)