Key Documents
GBL666384
Grace Bio-Labs Press-To-Seal silicone isolator, red silicone sheet
thickness 2.6 mm
Synonym(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
material
red silicon
sterility
non-sterile
feature
(Adhesive on both sides)
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 666384
L × W
180 mm × 130 mm
thickness
2.6 mm
General description
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)