Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GBL665217

Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side

wells, 16 (Square, 7mm x 7mm x 1.7mm depth), (Adhesive on one side)

Synonym(s):

Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

material

red silicon

sterility

non-sterile

feature

wells 16 (Square, 7mm x 7mm x 1.7mm depth)
(Adhesive on one side)

packaging

pack of 25 ea

manufacturer/tradename

Grace Bio-Labs 665217

external L × W

75 mm × 25 mm

well area

0.49 cm2

wells

16

Looking for similar products? Visit Product Comparison Guide

General description

Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.

Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service