Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GBL622510

Grace Bio-Labs SecureSeal hybridization chambers

wells, 16, well L × W × depth 7 mm × 7 mm × 0.8 mm, port diam. 1.5 mm

Synonym(s):

microarray chambers for slides, single use silicone-gasketed chamber

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

clear polycarbonate backing (0.25mm thickness)
red silicon chamber

sterility

non-sterile

feature

wells 16

packaging

pack of 25 ea

manufacturer/tradename

Grace Bio-Labs 622510

backing thickness

0.25 mm

external L × W

75 mm × 25 mm

port diam.

1.5 mm

well L × W × depth

7 mm × 7 mm × 0.8 mm

wells

16

Looking for similar products? Visit Product Comparison Guide

General description

SecureSeal Hybridization Chambers are thin, silicone-gasketed chambers providing optimal surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and microarrays on glass or film coated slides. These adhesive-backed chambers are designed for single-use, very-low volume incubations and are ideal for autoradiographic, fluorescent or chemiluminescent end-point applications requiring small, enclosed fluid volumes. The SecureSeal Adhesive is a double sided tape with a silicone adhesive surface on one side (covered with a clear PET liner) and an acrylic adhesive on the other surface (covered with a paper liner).

SecureSeal chambers form removable, peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. The chamber design minimizes friction, promotes reagent mixing, and facilitates uniform hybridization. Sealable access ports in the chamber surface allow for the addition and removal of reactants. SecureSeal adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating. Adhesive Seal Tabs create leak-proof chambers that are temperature resistant. Each package includes illustrated instructions for use and 120 adhesive port seals. Additional seal tabs are sold separately GBL629200). Press fit tubing connectors provide affordable and easy means to connect SecureSeal Hybridization Chambers to pumps and other devices for fluidic exchange.

Note: If you are using Cy5 of Alexa Fluor 647 direct-labeled DNA probes, please refer to "fluorescent friendly" chambers

Application

In situ hybridization
Protein and DNA Microarrays
Immunocytochemistry
DNA Amplification
Neurophysiology
Reaction kinetics
Rapid microfluidic prototyping
Nucleic Acid Programmable Protein Arrays
Fluorescence Resonance Energy Transfer (FRET)
Single molecule fluorescence microscopy

Legal Information

SecureSeal is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service