Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8166

Sigma-Aldrich

Glucagon-Like Peptide 2 -Arg human trifluoroacetate salt

>90% (HPLC), solid

Synonym(s):

GLP-2-Arg, Preproglucagon 126-159

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C171H266N48O56S
CAS Number:
Molecular Weight:
3922.29
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

Quality Level

Assay

>90% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GCG(2641)

Amino Acid Sequence

His-Ala-Asp-Gly-Ser-Phe-Ser-Asp-Glu-Met-Asn-Thr-Ile-Leu-Asp-Asn-Leu-Ala-Ala-Arg-Asp-Phe-Ile-Asn-Trp-Leu-Ile-Gln-Thr-Lys-Ile-Thr-Asp-Arg

General description

Glucagon-like peptide 2 (GLP-2) is a member of the pro-glucagon gene family which is released in small and large intestinal circulation by enteroendocrine L cells. This peptide is composed of 33 amino acids.

Application

Glucagon-Like Peptide 2 (GLP-2) -Arg human trifluoroacetate salt has been used for injection in chickens to determine the effects of GLP-2 on the weight and morphology of small intestines.

Biochem/physiol Actions

Glucagon-like peptide 2 (GLP-2) influences the functionality of gastrointestinal tract by participating in the feedback loop to the brain for the control of food intake. It also controls digestion and absorption, and has tropic effects post chemotherapy injury or surgical resection. This peptide is also responsible for epithelial cell proliferation of the small bowel. Studies in rats show that this peptide enhances the effects of the ileal-brake hormones, GLP-1 and peptide YY. It also negatively regulates gastrointestinal motility and secretion. Thus, it has potential as a therapeutic agent in short-bowel syndrome.
Glucagon-like peptide 2 is a neuropeptide that inhibits food intake when injected into the lateral ventricles of the brain and may mediate satiety. It is also a growth factor that stimulates proliferation and blocks apoptosis of intestinal epithelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Jeppesen et al.
Gastroenterology, 120(4), 806-815 (2001-03-07)
Glucagon-like peptide 2 (GLP-2) is intestinotrophic, antisecretory, and transit-modulating in rodents, and it is mainly secreted from the intestinal mucosa of the terminal ileum and colon after food ingestion. We assessed the effect of GLP-2 on the gastrointestinal function in
X F Hu et al.
Poultry science, 89(9), 1967-1974 (2010-08-17)
An experiment was conducted to determine the effect of injecting glucagon-like peptide 2 (GLP-2) on the small intestinal weight, morphology, and nutrient transporter expression in pharmacologically stressed broiler chickens. A total of 144 seven-day-old birds were fed either a basal
D J Drucker et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(15), 7911-7916 (1996-07-23)
Injury, inflammation, or resection of the small intestine results in severe compromise of intestinal function. Nevertheless, therapeutic strategies for enhancing growth and repair of the intestinal mucosal epithelium are currently not available. We demonstrate that nude mice bearing subcutaneous proglucagon-producing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service