Skip to Content
MilliporeSigma
All Photos(1)

Documents

G8035

Sigma-Aldrich

Anti-Glial Cell-derived Neurotrophic Factor (GDNF) antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GDNF(2668)

General description

The antibody neutralizes the biological activity of recombinant human GDNF.

Immunogen

recombinant human GDNF expressed in Escherichia coli.

Biochem/physiol Actions

Glial cell-derived neurotrophic factor is a potent neurotrophic factor that is important for the maintenance, regeneration and survival of specific neuronal cells including autonomic neurons, motoneurons, noradrenergic and seotonergic neurons. Additionally, it promotes the invasion and metastatic behavior of certain types of cancers by activating ERK, p38, JNK and AP-1 mediated cell signaling.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikko Airavaara et al.
Parkinsonism & related disorders, 18 Suppl 1, S143-S146 (2011-12-23)
Although initially thought to be important primarily in neural development, a number of trophic proteins have been found to have neuroprotective and neuroregenerative activity in the adult central nervous system, particularly for midbrain dopamine neurons (MDN). Neurorestoration is potentially feasible
Shelley J Allen et al.
Pharmacology & therapeutics, 138(2), 155-175 (2013-01-26)
Glial cell-derived neurotrophic factor (GDNF), and the neurotrophin nerve growth factor (NGF) and brain-derived neurotrophic factor (BDNF) are important for the survival, maintenance and regeneration of specific neuronal populations in the adult brain. Depletion of these neurotrophic factors has been
Jing-Yuan Chuang et al.
Oral oncology, 49(12), 1103-1112 (2013-09-28)
Perineural invasion is a prominent clinical feature of various cancers, which causes difficulty in curative resection. Glial cell-derived neurotrophic factor (GDNF), a potent neurotrophic factor, plays an important role in the invasive and metastatic behavior of various cancers. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service