Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

G7274

Sigma-Aldrich

DL-Glyceric acid hemicalcium salt hydrate

≥98% (GC)

Synonym(s):

2,3-Dihydroxypropanoic acid hemicalcium salt, Hemicalcium DL-glycerate, hemi-Calcium DL-glycerate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOCH2CH(OH)COO]2Ca · ×H2O
CAS Number:
Molecular Weight:
250.22 (anhydrous basis)
Beilstein:
4614104
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (GC)

form

crystalline

technique(s)

gas chromatography (GC): suitable

impurities

<18% water (Karl Fischer)

color

beige

mp

134 °C (dec.) (lit.)

solubility

50 mg/mL, clear, colorless to faintly yellow (1 N Hydrochloric acid)

cation traces

Ca: 15.5-16.5% (anhydrous)

SMILES string

[H]O[H].[H]O[H].OCC(O)C(=O)O[Ca]OC(=O)C(O)CO

InChI

1S/2C3H6O4.Ca.2H2O/c2*4-1-2(5)3(6)7;;;/h2*2,4-5H,1H2,(H,6,7);;2*1H2/q;;+2;;/p-2

InChI key

OKEVQKKVYQHNFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

DL-glyceric acid is also known as 2,3-dihydroxypropanoic acid.

Biochem/physiol Actions

Glyceric acid is an intermediate in the glycolysis pathway.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Drake et al.
Magnetic resonance in chemistry : MRC, 44(3), 210-219 (2006-02-16)
Vicinal (1)H--(1)H coupling constants were used to determine the conformational preferences of 2,3-dihydroxypropanoic acid (1) (DL-glyceric acid) in various solvents and its different carboxyl ionization states. The stereospecific assignments of J(12) and J(13) were confirmed through the point-group substitution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service