Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5798

Sigma-Aldrich

[10]-Gingerol

≥98% (HPLC)

Synonym(s):

(S)- 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone, 10-Gingerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CCCCCCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C21H34O4/c1-3-4-5-6-7-8-9-10-18(22)16-19(23)13-11-17-12-14-20(24)21(15-17)25-2/h12,14-15,18,22,24H,3-11,13,16H2,1-2H3/t18-/m0/s1

InChI key

AIULWNKTYPZYAN-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

[10]-Gingerol is a pungent, non-volatile phenolic compound of fresh ginger root.

Application

[10]-Gingerol has been used as an antibacterial agent to study its effects on Escherichia coli ATP synthase. It has also been used as an anti-hyperglycemic agent to test its effects on promoting glucose utilization in 3T3-L1 adipocytes and C2C12 myotubes.

Biochem/physiol Actions

[10]-Gingerol is a potent anti-cancer agent and a known inhibitor of breast cancer cells growth by blocking cell proliferation and inducing programmed cell death. It also inhibits ovarian cancer cells growth by inducing G2 phase cell cycle arrest. 10-Gingerol elicits inhibitory effects towards triple breast cancer cells in both mouse models and in vitro studies. It also displays anti-neuroinflammatory and anti-hyperglycemic activities.
[10]-Gingerol is a bioactive compound found in ginger (Zingiber officinale) with anti-inflammatory and antioxidant activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

331.3 °F - closed cup

Flash Point(C)

166.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Rasmussen et al.
Advanced pharmaceutical bulletin, 9(4), 685-689 (2019-12-21)
Purpose: Gingerol homologs found in the rhizomes of ginger plants have the potential to benefit human health, including the prevention and treatment of cancer. This study evaluated the effect of 10-gingerol on ovarian cancer cell (HEY, OVCAR3, and SKOV-3) growth.
Megan M Bernard et al.
Experimental and molecular pathology, 102(2), 370-376 (2017-03-21)
The ginger rhizome is rich in bioactive compounds, including [6]-gingerol, [8]-gingerol, and [10]-gingerol; however, to date, most research on the anti-cancer activities of gingerols have focused on [6]-gingerol. In this study, we compared [10]-gingerol with [8]-gingerol and [6]-gingerol in terms
Taurin Hughes et al.
International journal of biological macromolecules, 182, 2130-2143 (2021-06-05)
For centuries, dietary ginger has been known for its antioxidant, anticancer, and antibacterial properties. In the current study, we examined the link between antibacterial properties of 7 dietary ginger phenolics (DGPs)-gingerenone A, 6-gingerol, 8-gingerol, 10-gingerol, paradol, 6-shogaol, and zingerone-and inhibition
Su-Chen Ho et al.
Food chemistry, 141(3), 3183-3191 (2013-07-23)
Despite the anti-neuroinflammatory capacity of ginger, the corresponding active constituents are unclear. This study analyzed the composition of fresh ginger ethanolic extract by using LC-MS. Inhibitory activities of fresh ginger extract and seven gingerol-related compounds on the neuro-inflammation were also
Meran Keshawa Ediriweera et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-16)
10-gingerol is a major phenolic lipid found in the rhizomes of ginger (Zingiber officinale). Being amphiphilic in nature, phenolic lipids have the ability to incorporate into cell membranes and modulate membrane properties. The purpose of the present study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service