Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5547

Sigma-Aldrich

GSK837149A

≥98% (HPLC), solid

Synonym(s):

N,N′-Di[4-(4-Methyl-pyrimidin-2-ylsulfamoyl)phenyl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22N8O5S2
CAS Number:
Molecular Weight:
554.60
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >10 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

Biochem/physiol Actions

GSK837149A is a selective inhibitor of human fatty acid synthase (FAS).
GSK837149A is the first selective inhibitor of human fatty acid synthase (FAS) known to act specifically and selectively on the KR activity of the enzyme. It was first isolated as a minor impurity in a sample found to be active against the enzyme in a high-throughput screening campaign. This compound and its analogs synthesized, all being symmetrical structures containing a bisulfonamide urea, act by inhibiting the beta-ketoacyl reductase activity of the enzyme. GSK837149A inhibits FAS in a reversible mode, with a Ki value of approximately 30 nm, and it possibly binds to the enzyme-ketoacyl-ACP complex. Although initial results suggest that cell penetration for these compounds is impaired, they still can be regarded as useful tools with which to probe and explore the beta-ketoacyl reductase active site in FAS, helping in the design of new inhibitors.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David Weigt et al.
Cell chemical biology, 26(9), 1322-1331 (2019-07-08)
Human cancers require fatty acid synthase (FASN)-dependent de novo long-chain fatty acid synthesis for proliferation. FASN is therefore an attractive drug target, but fast technologies for reliable label-free cellular compound profiling are lacking. Recently, MALDI-mass spectrometry (MALDI-MS) has emerged as
Prosanta K Singha et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 149, 105321-105321 (2020-04-11)
De novo synthesis of fatty acids is essential to maintain intensive proliferation of cancer cells. Unlike normal cells that utilize food-derived circulating lipids for their fuel, cancer cells rely on heightened lipogenesis irrespective of exogenous lipid availability. Overexpression and activity

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service