Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5402

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–Gold antibody produced in rabbit

affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

gold conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

aqueous glycerol suspension

technique(s)

dot blot: suitable

particle size

10 nm (colloidal gold)

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Primary goat antibodies are often used to study target proteins for various clinical and research purposes. Thus, secondary antibodies against goat IgGs can be used to facilitate the accurate detection and localization of target proteins. Anti-Goat IgG (whole molecule)-Gold antibody binds all goat immunoglobulins.

Immunogen

Purified goat IgG

Application

Anti-Goat IgG (whole molecule)-Gold antibody is suitable for use in immunoelectron microscopy. The antibody can also be used for dot blot.

Physical form

Colloidal suspension in Tris buffered saline, pH 8.2, with 30% glycerol (v/v), 1% bovine serum albumin (w/v) and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujiang Shang et al.
The Journal of cell biology, 215(3), 369-381 (2016-11-02)
Transient receptor potential A1 (TRPA1) is a nonselective cation channel implicated in thermosensation and inflammatory pain. In this study, we show that TRPA1 (activated by allyl isothiocyanate, acrolein, and 4-hydroxynonenal) elevates the intracellular Ca2+ concentration ([Ca2+]i) in dorsal root ganglion
L Moysset et al.
Planta, 213(4), 565-574 (2001-09-15)
The intracellular localization of phytochrome in the pulvini of Robinia pseudoacacia L. was analyzed by immunogold electron microscopy after red (R; 15 min) and far-red (FR; 5 min) irradiation 2 h after the beginning of the photoperiod. Screening of the
Characterization of Immunological Interactions at an Immunoelectrode by Scanning Electron Microscopy
Ambrosi, A., et al.
Electroanalysis, 19(2-3), 244-252 (2007)
Zhicheng Peng et al.
Frontiers in immunology, 12, 726829-726829 (2021-10-26)
β2 integrins are critical for neutrophil firm adhesion, trans-endothelial migration, and the recruitment to the inflamed tissue. Autophagy is implicated in cell migration and tumor metastasis through facilitating the turnover of β1 integrins; however, whether autophagy is able to control
Elie Akoury et al.
Human reproduction (Oxford, England), 30(1), 159-169 (2014-11-02)
What is the subcellular localization in human oocytes and preimplantation embryos, of the two maternal-effect proteins, NLRP7 and KHDC3L, responsible for recurrent hydatidiform moles (RHMs)? NLRP7 and KHDC3L localize to the oocyte cytoskeleton and are polar and absent from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service