Skip to Content
MilliporeSigma
All Photos(1)

Documents

G5261

Sigma-Aldrich

Gly-Asp-Asp-Asp-Asp-Lys-β-naphthylamide

≥95% (HPLC)

Synonym(s):

H-Gly-Asp-Asp-Asp-Asp-Lys-bNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H44N8O14
CAS Number:
Molecular Weight:
788.76
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NCCCCC(NC(=O)C(CC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC(O)=O)NC(=O)CN)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C34H44N8O14/c35-10-4-3-7-20(30(52)37-19-9-8-17-5-1-2-6-18(17)11-19)39-32(54)22(13-27(46)47)41-34(56)24(15-29(50)51)42-33(55)23(14-28(48)49)40-31(53)21(12-26(44)45)38-25(43)16-36/h1-2,5-6,8-9,11,20-24H,3-4,7,10,12-16,35-36H2,(H,37,52)(H,38,43)(H,39,54)(H,40,53)(H,41,56)(H,42,55)(H,44,45)(H,46,47)(H,48,49)(H,50,51)

InChI key

QLLWULMEPVZWTP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

A sensitive and specific fluorogenic substrate for enterokinase.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Antonowicz et al.
Clinica chimica acta; international journal of clinical chemistry, 101(1), 69-76 (1980-02-14)
The application of a new synthetic substrate to the direct determination of enteropeptidase is described. The substrate Gly-(L-Asp)4-L-Lys-2-naphthylamide contains the amino acid sequence of the activation peptides of trypsinogen linked via an amide bond to the fluorophore 2-naphthylamine. The sequence
Weimei Sun et al.
The Journal of pharmacology and experimental therapeutics, 375(3), 510-521 (2020-10-10)
Inhibition of the serine protease enteropeptidase (EP) opens a new avenue to the discovery of chemotherapeutics for the treatment of metabolic diseases. Camostat has been used clinically for treating chronic pancreatitis in Japan; however, the mechanistic basis of the observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service