Skip to Content
MilliporeSigma

G3002

Gly-Pro

≥98%, suitable for ligand binding assays

Synonym(s):

glycyl-L-proline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H12N2O3
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
EC Number:
211-880-2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Gly-Pro,

assay

≥98%

Quality Segment

form

solid

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

NCC(N1[C@H](C(O)=O)CCC1)=O

InChI

1S/C7H12N2O3/c8-4-6(10)9-3-1-2-5(9)7(11)12/h5H,1-4,8H2,(H,11,12)

InChI key

KZNQNBZMBZJQJO-UHFFFAOYSA-N

Application


  • Diagnostic efficacy of [(68)Ga]Ga-DOTA-GPFAPI-04 in patients with solid tumors in a head-to-head comparison with [(18)F]F-FDG: results from a prospective clinical study.: This study explores the use of Gly-Pro in peptide imaging agents, specifically [(68)Ga]Ga-DOTA-GPFAPI-04, demonstrating its superior diagnostic performance in detecting solid tumors compared to traditional imaging modalities, offering new avenues for cancer diagnosis and monitoring (Yuan et al., 2024).

  • Understanding the Transepithelial Transport and Transbilayer Diffusion of the Antihypertensive Peptide Asn-Cys-Trp: Insights from Caco-2 Cell Monolayers and the DPPC Model Membrane.: This research demonstrates the utility of Gly-Pro in studying the absorption and metabolic stability of antihypertensive peptides, providing key insights into their pharmacokinetic behaviors and enhancing drug delivery strategies (Wu et al., 2024).



pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library