Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G111040

Sigma-Aldrich

DMT-dG(ib) Phosphoramidite

configured for MerMade

Synonym(s):

DMT-dG(ib) amidite, Guanosine, 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-N-(2-methyl-1-oxopropyl)-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H54N7O8P
CAS Number:
Molecular Weight:
839.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)

color

white to off-white

λ

conforms (UV/VIS Identity)

compatibility

configured for MerMade

nucleoside profile

base: deoxyguanosine
base protecting group: isobutyryl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C44H54N7O8P/c1-28(2)41(52)48-43-47-40-39(42(53)49-43)46-27-50(40)38-25-36(59-60(57-24-12-23-45)51(29(3)4)30(5)6)37(58-38)26-56-44(31-13-10-9-11-14-31,32-15-19-34(54-7)20-16-32)33-17-21-35(55-8)22-18-33/h9-11,13-22,27-30,36-38H,12,24-26H2,1-8H3,(H2,47,48,49,52,53)/t36-,37+,38+,60?/m0/s1

InChI key

FDRMKYVTIFSDPR-MMROLVBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proligo′s DNA phosphoramidites lead to high-yield and high-quality oligonucleotides due to their high coupling efficiency. The deprotection step of automated oligonucleotide synthesis is integral to synthesis time and final product quality. Exocyclic amine functions are protected by a benzoyl group (dA(bz) and dC(bz)) or isobutyryl group (dG(ib))Key features of DNA phosphoramidites:
  • Recommended cleavage and deprotection conditions are 8 hours at 55 °Cor 24 hours at room temperature using concentrated ammonia solution, for standard base-protected oligonucleotides
  • The high coupling efficiency of Proligo′s DNA phosphoramidites leads to high-yield and high-quality oligonucleotides
The products belong to MerMade synthesizers.

Packaging

G111040-1G packaged in a 1 oz bottle with a 28-400 cap
G111040-10G packaged in a 8 oz bottle with a 28-400 cap
G111040-6x5G packaged in a 8 oz bottle with a 28-400 cap
G111040-6x10G packaged in a 8 oz bottle with a 28-400 cap
G111040-20x10G packaged in a 1 L bottle with GL 45

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service