Skip to Content
MilliporeSigma

FG1385

RPMI-1640 Medium

HEPES modification, with stable glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES and stable glutamine, liquid, sterile-filtered

Quality Segment

sterility

sterile-filtered, sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

components

stable glutamine: yes
phenol red: yes
HEPES: 25 mM
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Application

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells.


Still not finding the right product?

Explore all of our products under RPMI-1640 Medium


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library