Skip to Content
MilliporeSigma
All Photos(1)

Documents

FG0815

Sigma-Aldrich

Nutrient Mixture F-12 Ham

With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Nutrient Mixture F-12 Ham, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

stable glutamine: yes
HEPES: no
sodium pyruvate: 0.11 g/L
phenol red: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service