Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

F9634

Sigma-Aldrich

DL-Fluorocitric acid barium salt

Synonym(s):

Barium Fluorocitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H8F2O14Ba3
CAS Number:
Molecular Weight:
826.16
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

impurities

≤5% acetic acid

Quality Level

SMILES string

OC(CC([O-])=O)(C(C([O-])=O)F)C([O-])=O.OC(CC([O-])=O)(C(C([O-])=O)F)C([O-])=O

InChI

1S/C6H7FO7.Ba.2H/c7-3(4(10)11)6(14,5(12)13)1-2(8)9;;;/h3,14H,1H2,(H,8,9)(H,10,11)(H,12,13);;;

InChI key

LHKWNDHJHASTSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Fluorocitric acid barium salt has been used as an astrocyte inhibitor in various experiments.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of chemokine CXCL 11 in the development of morphine tolerance in rats with cancer-induced bone pain
Guo G, et al.
Journal of Neurochemistry, 141(4), 553-564 (2017)
Olga S Belozor et al.
Brain sciences, 9(4) (2019-04-25)
Astrogliosis is a pathological process that affects the density, morphology, and function of astrocytes. It is a common feature of brain trauma, autoimmune diseases, and neurodegeneration including spinocerebellar ataxia type 1 (SCA1), a poorly understood neurodegenerative disease. S100β is a
Astrocyte contributes to pain development via MMP2-JNK1/2 signaling in a mouse model of complex regional pain syndrome
Tian G, et al.
Life Sciences, 170, 64-71 (2017)
CXCL12/CXCR4 signaling contributes to neuropathic pain via central sensitization mechanisms in a rat spinal nerve ligation model
Liu ZY, et al.
CNS Neuroscience & Therapeutics (2019)
Muhammad Tahir Khan et al.
Brain research bulletin, 151, 164-173 (2018-08-12)
Whole-cell patch clamp recordings demonstrated that in the dentate gyrus (DG) as well as in the CA3 area of mouse hippocampal slices the prototypic P2X7 receptor (R) agonist dibenzoyl-ATP (Bz-ATP) induced inward current responses both in neurons and astrocytes. Whereas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service