Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F9307

Sigma-Aldrich

FSL-A(tri)

Synonym(s):

FSL-A(GALNa3[Fa2]GALb)-SA1-L1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H125N3NaO25P
Molecular Weight:
1462.71
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

≥98% (TLC)

Quality Level

form

solid

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

Application

All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. Cells modified with KODE constructs are known as kodecytes and usually maintain their normal vitality and functionality.

FSL-A(tri) has been specifically designed to create live cells (kodecytes) expressing blood group A antigens but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces. This product can be used to attach reproducible and controlled levels of blood group A antigens (generic A due to its trisaccharide structure) to group O or B erythrocytes. These kodecytes will react with most anti-A blood grouping reagents including polyclonal reagents

Other Notes

FSL-A(tri) is a KODE technology construct designed to label hydrophobic surfaces including living cells with blood group A trisaccharide. All KODE FSL constructs consist of three essential designable features: a functional component (F), a spacer (S), and a diacyl lipid (L). FSL-A(tri) is comprised of the blood group A trisaccharide GalNAcα3(Fucα2)Galβ representing F, conjugated via an O(CH2)3NH spacer (SA1) to an activated adipate derivative of dioleoyl­phospha­tidyl­ethanolamine (L1).

Legal Information

KODE Constructs are supplied by KODE Biotech Materials Limited under license from KODE Biotech Limited.
KODE is a trademark of KODE Biotech Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service