MilliporeSigma
Skip To
F8145
Carestream® electron image film SO-163
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
MA.01
UNSPSC Code:
41105341
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sheet size
3 1/4 in. × 4 in.
storage temp.
2-8°C
General description
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Greater electron exposure and development latitudes than 4489.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)