Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥1800 units/μg protein
Assay:
≥95% (SDS-PAGE)
Recombinant:
expressed in Sf21 cells
recombinant
expressed in Sf21 cells
Quality Level
assay
≥95% (SDS-PAGE)
form
solution
specific activity
≥1800 units/μg protein
impurities
≤1.0 EU/μg endotoxin
shipped in
dry ice
storage temp.
−70°C
Related Categories
General description
Fibroblast activation protein alpha (FAPa) is a type II transmembrane serine protease. It is present as a homodimeric integral protein with dipeptidyl peptidase IV like fold. It has an α/β-hydrolase domain and an eight-bladed β-propeller domain. It is not expressed in normal tissue. It is only expressed by activated fibroblast in response to pathologic situations. It is expressed in several pathogenic sites including cancer, fibrosis, arthritis, wounding, or inflammation.
Application
FAPa may be used to study the role in coordinating the stromal response to cancer, in angiogenesis, and modulation of the antitumor immune response.
Biochem/physiol Actions
FAP is a Type II transmembrane serine protease which has substrate specificity for prolyl peptides bonds (Pro-Xaa). This endopeptidase cleaves gelatin and is believed to play a role in tissue remodeling, tumor invasion, and wound repair.
FAPα has in vitro dipeptidyl peptidase activity and collagenolytic activity. It cleaves N-terminal dipeptides from polypeptides and can degrade gelatin and type I collagen. It has also been reported that FAPa has a tumor suppressor activity.
Physical form
Supplied as a solution in 25 mM Tris, 0.25 M NaCl and 20% (v/v) glycerol , pH 8.0
Other Notes
One unit will convert 1 picomole of benzyloxycarbonyl-Gly-Pro-7-amido-4-methylcoumarin (ZP-AMC) to Z-gly-Pro and 7-amino-4-methylcoumarin (AMC) per minute at pH 7.5 at 25°C.
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kathleen Aertgeerts et al.
The Journal of biological chemistry, 280(20), 19441-19444 (2005-04-06)
Fibroblast activation protein alpha (FAPalpha) is highly expressed in epithelial cancers and has been implicated in extracellular matrix remodeling, tumor growth, and metastasis. We present the first high resolution structure for the apoenzyme as well as kinetic data toward small
Thomas Kelly et al.
International review of cell and molecular biology, 297, 83-116 (2012-05-23)
Fibroblast activation protein-α (FAP) is a serine protease that can provide target specificity to therapeutic agents because in adults its expression is restricted to pathologic sites, including cancer, fibrosis, arthritis, wounding, or inflammation. It is not expressed in most normal
Dimitrios Patsouras et al.
Molecular medicine reports, 11(6), 4585-4590 (2015-01-28)
Fibroblast activation protein (FAP), a selective protein for tumor stromal fibroblasts, is expressed in >90% of human epithelial carcinomas. A characteristic feature of pancreatic cancer is an extensive fibrotic or desmoplastic reaction surrounding the primary tumor. The present study aimed
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F7182-10UG | 04061832700786 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service