Skip to Content
MilliporeSigma

F2036

Nalgene® filtration assembly for 47 mm filter

autoclavable

Synonym(s):

filtration device, flask top filter, Nalgene DS0315-0047, filter, liquid sieve

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polysulfone housing

feature

autoclavable

packaging

pack of 1 ea

manufacturer/tradename

Nalgene DS0315-0047

funnel capacity

250 mL

neck diam.

33-41 mm

application(s)

filtration

General description

Assembly consists of a polysulfone funnel (250 mL capacity), polypropylene copolymer cover with three ports, thermoplastic elastomer cap, silicone gasket, #8 rubber stopper, and aluminum clamp. Can be used with any standard filter flask or filter manifold.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library