Skip to Content
MilliporeSigma

F1672

Anti-Fibroblast Growth Factor-9 antibody, Mouse monoclonal

clone FG9-77, purified from hybridoma cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
FG9-77, monoclonal
Application:
ARR, DB, ELISA (i), WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FG9-77, monoclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

species reactivity

mouse, human

technique(s)

dot blot: 5 μg/mL using recombinant human FGF-9, indirect ELISA: suitable, microarray: suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF9(2254)
mouse ... Fgf9(14180)

General description

Fibroblast Growth Factor-9 (FGF-9) also refers as glia activating factor is a glycosylated single polypeptide of 208 amino acids that belongs to heparin-binding growth factors family. It has a crucial role in the proliferation of oligodendrocyte type 2 astrocyte progenitor cells, rat pheochromacytoma cell line PC-12 and Balb/c 3T3 fibroblasts. It mainly expresses in brain and kidney. Monoclonal Anti-Fibroblast Growth Factor-9 antibody can be used for studying the fibroblast growth factors and their properties as well as to compare its expression pattern with physiological functions or pathological conditions. It can also be used in immunoblotting.

Immunogen

recombinant mouse FGF-9.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Monoclonal Anti-Fibroblast Growth Factor-9 antibody can be used in several immunochemical assays like dot blot and ELISA for the localization of FGF-9. It can also be used in microarray.

Biochem/physiol Actions

The antibody reacts specifically with recombinant FGF-9. It does not cross-react with human acidic FGF or basic FGF. Mouse anti-Fibroblast Growth Factor-9 antibody reacts specifically with recombinant FGF-9.

Physical form

Solution, 0.2 μm filtered, in 0.01 M phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library