Documents
EPM1289-0002
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
size small, holds 34 tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.73
size
small
300x80x129 mm
feature
holds 34 tubes
manufacturer/tradename
Eppendorf® M1289-0002 (Global)
Related Categories
General description
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG