Skip to Content
MilliporeSigma
All Photos(1)

Documents

EP6313000018

Eppendorf® Mastercycler®

X50a, with control panel, aluminum block, size 96 wells, AC/DC input 110 V / 230 V, 50-60 Hz (NA/EU)

Synonym(s):

dry bath, heat block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NC.21

material

aluminum block

feature

Unit can control 9 other units in network
holds 96×0.2 mL PCR vessels, 1 PCR plate 8 × 12
unit can only be controlled
with control panel

manufacturer/tradename

Eppendorf® 6313000018

parameter

1-30 °C (gradient range)
30-99 °C (gradient temperature)
37-110 °C temp. range
4-99 °C temp. range (control range)

technique(s)

PCR: suitable

W × D × H

27.5 cm × 43 cm × 33 cm

size

96 wells

weight

11.5 kg

Looking for similar products? Visit Product Comparison Guide

General description

The Eppendorf Mastercycler X50 Thermocycler combines speed, flexibility and PCR optimization for research PCR applications and standardization for routine PCR applications such as food testing. A highly intuitive touch screen puts all these benefits at your fingertips.

Speed and enhanced PCR optimization functions, like the 2D-Gradient, make the Eppendorf Mastercycler X50 Thermocycler the ideal PCR cycler for advanced research in molecular biology. The excellent block temperature regulation gives rise to the next stage of PCR reproducibility, whereas the adaptable user management and profound documentation capabilities give peace of mind to laboratories working to specific or strict standards.

A highly intuitive touch display, low noise levels, low power consumption and the versatile flexlid concept combine to make a powerful PCR cycler. Up to 10 PCR cyclers can be combined – ideal for high throughput applications or labs with a high number of users running different assays. Should you feel you need more flexibility or throughput, up to 50 PCR cyclers can be combined in a computer-controlled network.

Product Highlights:
PCR optimization with 2D-Gradient

Denaturation temperature gradient (top to bottom by row)
Annealing temperature gradient (left to right by column)

Faster Heating and Cooling

Heating rate: up to 10°C/ sec. Eppendorf Mastercycler x50 Thermocycler
Cooling rate: 5°C/ sec.
Lid Height Adjusts Automatically

flexlid concept- the automatically adjusts to the heights of plates and tubes; allowing you to use all types of consumables

Color Touchscreen

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service