Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5820733004

Eppendorf® Adapter, for Eppendorf Conical Tubes with snap cap

tube capacity 25 mL, case of 6 ea, for use with for centrifuges with rotors for conical 50 mL tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.01

packaging

case of 6 ea

manufacturer/tradename

Eppendorf® 5820733004

tube capacity

25 mL

compatibility

for use with for centrifuges with rotors for conical 50 mL tubes

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service