Key Documents
EP4982000322
Eppendorf® Repeater M4
starter kit with Combitip Rack, with Combitip assortment pack, 1 ul - 10 ml
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
volume:
1—10000 μL
sterility:
non-sterile
feature:
starter kit with Combitip Rack
with Combitip assortment pack
packaging:
pack of 1 ea (starter kit with Combitip Rack)
manufacturer/tradename:
Eppendorf® 4982000322 (US)
sterility
non-sterile
feature
starter kit with Combitip Rack
with Combitip assortment pack
packaging
pack of 1 ea (starter kit with Combitip Rack)
manufacturer/tradename
Eppendorf® 4982000322 (US)
measuring range
1 ul - 10 ml
maximum volume
10 mL
volume
1—10000 μL
General description
With the Repeater M4/Combitip System, volumes are dispensed using the positive displacement principle. The liquid is directly dispensed without an air cushion, ensuring the correct volume is always dispensed regardless of the density, viscosity and volatility of the liquid. The Repeater M4 offers a unique volume range of 1 μL – 10 mL, and is the only mechanical dispenser that dispenses the aspirated liquid up to 100 times without refilling the tip. The innovative step counter supports error-free working even after an interruption or distraction during the dispensing procedure. A fully emptied Combitip can be easily ejected with one hand using the operation lever without directly touching the tip.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)