Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP4861000724

Eppendorf® Xplorer® Plus single channel pipettor

volume 15-300 μL, multifunction rocker switch, orange, for use with for pipette tips 300 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

feature

multifunction rocker switch

packaging

case of 1 ea

manufacturer/tradename

Eppendorf® 4861000724

volume

15-300 μL

color

orange

compatibility

for use with for pipette tips 300 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Multi-functional rocker with up is up and down is down functionality
  • Selection dial for quick and easy selection of functions
  • Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
  • Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
  • Powerful rechargeable battery for up to 8 hours work without the need for recharging
  • Separate power socket: Work can continue while charging
  • Adjustment to external conditions (e.g. geographic height, liquids with high density)
  • Lower part of pipette can be autoclaved to ensure decontamination
  • 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Xplorer is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service