Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP3125000206

Eppendorf® Research® Plus Move It pipette

no. of channels, 8, volume 120-1200 μL, cone distance 4.5-14 mm, dark green

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.51

material

dark green

feature

no. of channels 8

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 3125000206

cone distance

4.5-14 mm

volume

120-1200 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Adjustable tip spacing pipette
  • Multi-channel pipette
  • 8-channel
  • 120-1200μL
  • 9mm cone distance
  • Dark green

Application

  • Forward pipetting
  • Reverse pipetting
  • Removal of supernatants
  • Sample mixing
  • Phase extraction
  • Filling of plates, gels, and reaction vessel

Features and Benefits

Air cushioned pipettes with variable volume and adjustable tip spacing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service