Key Documents
EP022639200
Eppendorf® Adapter
to hold, 2 x 15mL, for 100 mL round bucket in Rotor A-4-38, pack of 2 ea
About This Item
UNSPSC Code:
41103911
material
POM
feature
to hold 2 x 15mL, for 100 mL round bucket in Rotor A-4-38
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022639200
compatibility
Rotor A-4-38
Related Categories
General description
Adapter, for 2 conical tubes 15 mL, 17.2 mm × 121 mm, for 100 mL round bucket in Rotor A-4-38, 2 pcs.
  • for 100 mL round bucket in Rotor A-4-38
  • Please note that this adapter is not compatible with the aerosol-tight caps of this rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)