Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU101391

Sigma-Aldrich

MISSION® esiRNA

targeting human OPA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGTTGTTGTGGTTGGAGATCAGAGTGCTGGAAAGACTAGTGTGTTGGAAATGATTGCCCAAGCTCGAATATTCCCAAGAGGATCTGGGGAGATGATGACACGTTCTCCAGTTAAGGTGACTCTGAGTGAAGGTCCTCACCATGTGGCCCTATTTAAAGATAGTTCTCGGGAGTTTGATCTTACCAAAGAAGAAGATCTTGCAGCATTAAGACATGAAATAGAACTTCGAATGAGGAAAAATGTGAAAGAAGGCTGTACCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucie Potuckova et al.
Frontiers in immunology, 9, 1771-1771 (2018-08-18)
C-terminal Src kinase (CSK) is a major negative regulator of Src family tyrosine kinases (SFKs) that play critical roles in immunoreceptor signaling. CSK is brought in contiguity to the plasma membrane-bound SFKs via binding to transmembrane adaptor PAG, also known
Kamariah Ibrahim et al.
International journal of molecular medicine, 46(2), 685-699 (2020-05-30)
Glioblastoma multiforme (GBM) is an aggressive type of brain tumour that commonly exhibits resistance to treatment. The tumour is highly heterogenous and complex kinomic alterations have been reported leading to dysregulation of signalling pathways. The present study aimed to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service