Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU093011

Sigma-Aldrich

MISSION® esiRNA

targeting human TPX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCACCAAAGAAGATGAGGAAGAGGACGAACCGGTAGTGATAAAAGCTCAACCTGTGCCACATTATGGGGTGCCTTTTAAGCCCCAAATCCCAGAGGCAAGAACTGTGGAAATATGCCCTTTCTCGTTTGATTCTCGAGACAAAGAACGTCAGTTACAGAAGGAGAAGAAAATAAAAGAACTGCAGAAAGGGGAGGTGCCCAAGTTCAAGGCACTTCCCTTGCCTCATTTTGACACCATTAACCTGCCAGAGAAGAAGGTAAAGAATGTGACCCAGATTGAACCTTTCTGCTTGGAGACTGACAGAAGAGGTGCTCTGAAGGCACAGACTTGGAAGCACCAGCTGGAAGAAGAACTGAGACAGCAGAAAGAAGCAGCTTGTTTCAAGGCTCGTCCAAACACCGTCATCTCTCAGGAGCCCTTTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingquan Liu et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 45(8), 906-918 (2014-09-30)
Targeting protein for Xenopus kinesin-like protein 2 (TPX2) is a microtubule-associated protein that impacts spindle assembly in human cells. Several studies have shown that the overexpression of TPX2 is correlated with multiple tumor types. However, the role of TPX2 in
Tomohiro Miwa et al.
Cancer medicine, 4(7), 1091-1100 (2015-04-29)
The targeting protein for Xklp2 (TPX2) is a microtubule- and, cell cycle-associated protein who's overexpression has been reported in various malignancies. In this study, we verified the overexpression of TPX2 in both surgically resected specimens of pancreatic cancer and multiple
Yong Yang et al.
Asian Pacific journal of tropical medicine, 8(12), 1064-1070 (2015-12-27)
To investigate the expression of targeting protein for Xenopus kinesin-like protein 2 (TPX2) in breast cancer tissue and to explore its role in proliferation, migration and invasion of breast cancer cells. The mRNA and protein expressions of TPX2 in breast
Helen Chen et al.
Cell cycle (Georgetown, Tex.), 13(14), 2248-2261 (2014-05-31)
Construction of a mitotic spindle requires biochemical pathways to assemble spindle microtubules and structural proteins to organize these microtubules into a bipolar array. Through a complex with dynein, the receptor for hyaluronan-mediated motility (RHAMM) cross-links mitotic microtubules to provide structural
Yuqi Huang et al.
International journal of molecular sciences, 15(10), 18148-18161 (2014-10-11)
Targeting protein for Xenopus kinesin-like protein 2 (TPX2), a microtubule-associated protein, impacts spindle assembly in human cells. Several studies have demonstrated that TPX2 is overexpressed in different types of human cancers and promotes tumor growth and metastasis. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service