Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU032191

Sigma-Aldrich

MISSION® esiRNA

targeting human GSDMD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTTCTGGAAACCCCGTTATAAGTGTGTCAACCTGTCTATCAAGGACATCCTGGAGCCGGATGCCGCGGAACCAGACGTGCAGCGTGGCAGGAGCTTCCACTTCTACGATGCCATGGATGGGCAGATACAGGGCAGCGTGGAGCTGGCAGCCCCAGGACAGGCAAAGATCGCAGGCGGGGCCGCGGTGTCTGACAGCTCCAGCACCTCAATGAATGTGTACTCGCTGAGTGTGGACCCTAACACCTGGCAGACTCTGCTCCATGAGAGGCACCTGCGGCAGCCAGAACACAAAGTCCTGCAGCAGCTGCGCAGCCGCGGGGACAACGTGTACGTGGTGACTGAGGTGCTGCAGACACAGAAGGAGGTGGAAGTCACGCGCACCCACAAGCGGGAGGGCTCGGGCCGGTTTTCCCTGCCCGGAGCCACGTGCTTGCAGGGTGAGGGCCAGGGCCATCTGAGCCAGAAGAAGACGGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianwei Gao et al.
Oncology reports, 40(4), 1971-1984 (2018-08-15)
Gasdermin D (GSDMD) is a newly discovered pyroptosis executive protein, which can be cleaved by inflammatory caspases and is essential for secretion of IL‑1β, making it a critical mediator of inflammation. However, the precise role of GSDMD in carcinogenesis remains nearly
Hui Chen et al.
Molecular neurodegeneration, 15(1), 26-26 (2020-04-17)
Acute glaucoma, characterized by a sudden elevation in intraocular pressure (IOP) and retinal ganglion cells (RGCs) death, is a major cause of irreversible blindness worldwide that lacks approved effective therapies, validated treatment targets and clear molecular mechanisms. We sought to
Wenyi Zhao et al.
Experimental eye research, 202, 108375-108375 (2020-12-07)
The protein GSDMD is an important performer of pyroptosis and a universal substrate for the inflammatory caspase. However, the role and regulatory mechanism of GSDMD in Aspergillus fumigatus keratitis is remains unknown. Here we detected GSDMD protein in the cornea
Guangxue Gu et al.
International immunopharmacology, 91, 107227-107227 (2020-12-29)
Ankylosing spondylitis (AS) is a disease characterized by inflammation of the sacroiliac joint and the attachment point of the spine. This study aimed to investigate the effect of microRNA (miR)-204-targeted GSDMD on fibroblast-like synoviocytes (FLSs) in AS. miR-204, GSDMD, pyrolysis-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service