Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU026851

Sigma-Aldrich

MISSION® esiRNA

targeting human TBC1D9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGTGGACCAAGGTGTCTTTGAGGAGCTAGCACGAGACTACGTCCCACAGCTGTACGACTGCATGCAAGACCTGGGCGTGATTTCCACCATCTCCCTGTCTTGGTTCCTCACACTATTTCTCAGTGTGATGCCTTTTGAGAGTGCAGTTGTGGTTGTTGACTGTTTCTTCTATGAAGGAATTAAAGTGATATTCCAGTTGGCCCTAGCTGTGCTGGATGCAAATGTGGACAAACTGTTGAACTGCAAGGATGATGGGGAGGCCATGACCGTTTTGGGAAGGTATTTAGACAGTGTGACCAATAAAGACAGCACACTGCCTCCCATTCCTCACCTCCACTCCTTGCTCAGCGATGATGTGGAACCTTACCCTGAGGTAGACATCTTTAGACTCATCAGAACTTCCTACGAGAAATTCGGAACTATCCGGGCAGATTTGATTGAACAGATGAGATTCAAACAGAGACTGAAAGTGATCCAGACGCTG

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
Theranostics, 5(12), 1456-1472 (2015-12-19)
Understanding the molecular basis of drug resistance and utilising this information to overcome chemoresistance remains a key challenge in oncology. Here we report that survivin, a key protein implicated in drug resistance, is overexpressed in cancer stem cell pool of
Xiaoqian Yang et al.
Pharmaceutical research, 32(6), 2097-2109 (2014-12-18)
Approaches for the synthesis of biomaterials to facilitate the delivery of "biologics" is a major area of research in cancer therapy. Here we designed and characterized a hyaluronic acid (HA) based self-assembling nanoparticles that can target CD44 receptors overexpressed on
Runbi Ji et al.
Cell cycle (Georgetown, Tex.), 14(15), 2473-2483 (2015-06-20)
Mesenchymal stem cells (MSCs) play an important role in chemoresistance. Exosomes have been reported to modify cellular phenotype and function by mediating cell-cell communication. In this study, we aimed to investigate whether exosomes derived from MSCs (MSC-exosomes) are involved in
Takashi Nozawa et al.
Nature communications, 11(1), 770-770 (2020-02-09)
Invading microbial pathogens can be eliminated selectively by xenophagy. Ubiquitin-mediated autophagy receptors are phosphorylated by TANK-binding kinase 1 (TBK1) and recruited to ubiquitinated bacteria to facilitate autophagosome formation during xenophagy, but the molecular mechanism underlying TBK1 activation in response to
Xia Wen et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(2), 475-483 (2014-07-13)
Paraquat is a herbicide that is highly toxic to the lungs and kidneys following acute exposures. Prior studies have demonstrated that the organic cation transporter 2 and multidrug and toxin extrusion protein 1 contribute to the urinary secretion of paraquat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service