Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU016451

Sigma-Aldrich

MISSION® esiRNA

targeting human PTPN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TATCCGACATGAAGCCAGTGACTTCCCATGTAGAGTGGCCAAGCTTCCTAAGAACAAAAACCGAAATAGGTACAGAGACGTCAGTCCCTTTGACCATAGTCGGATTAAACTACATCAAGAAGATAATGACTATATCAACGCTAGTTTGATAAAAATGGAAGAAGCCCAAAGGAGTTACATTCTTACCCAGGGCCCTTTGCCTAACACATGCGGTCACTTTTGGGAGATGGTGTGGGAGCAGAAAAGCAGGGGTGTCGTCATGCTCAACAGAGTGATGGAGAAAGGTTCGTTAAAATGCGCACAATACTGGCCACAAAAAGAAGAAAAAGAGATGATCTTTGAAGACACAAATTTGAAATTAACATTGATCTCTGAAGATATCAAGTCATATTATACAGTGCGACAGCTAGAATTGGAAAACCTTACAACCCAAGAAACTCGAGAGATCTTACATTTCCACTATACCACATGGCCTGACTTTGGAGTCCCTGAATCACCAGCCTCATT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akarawin Hongdusit et al.
Nature communications, 11(1), 788-788 (2020-02-09)
Protein tyrosine phosphatases regulate a myriad of essential subcellular signaling events, yet they remain difficult to study in their native biophysical context. Here we develop a minimally disruptive optical approach to control protein tyrosine phosphatase 1B (PTP1B)-an important regulator of
Caroline E Nunes-Xavier et al.
Diagnostic pathology, 14(1), 134-134 (2019-12-16)
Protein tyrosine phosphatases (PTPs) regulate neuronal differentiation and survival, but their expression patterns and functions in human neuroblastoma (NB) are scarcely known. Here, we have investigated the function and expression of the non-receptor PTPN1 on human NB cell lines and
Liza D Morales et al.
PloS one, 9(5), e97104-e97104 (2014-05-09)
To maintain tissue homeostasis, apoptosis is functionally linked to the cell cycle through the retinoblastoma (Rb)/E2F pathway. When the Rb tumor suppressor protein is functionally inactivated, E2F1 elicits an apoptotic response through both intrinsic (caspase-9 mediated) and extrinsic (caspase-8 mediated)
Samuel Legeay et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 127, 110200-110200 (2020-05-18)
Diabetes notably increases the risk for endothelial dysfunction, a main precursor for microvascular complications. While endoplasmic reticulum stress (ERS) and protein tyrosine phosphatase 1B (PTP1B) have been associated with endothelial dysfunction in resistance vessels, whether these mechanisms also contribute to
Xiaoyan Zhang et al.
Scientific reports, 5, 12987-12987 (2015-08-12)
Renal mesangial cells (RMCs) constitute a population of cells in glomerular mesangium. Inflammatory cytokines produced by RMCs play a vital role in renal inflammation. miRNAs are key regulators of inflammatory cytokine expression. The abnormal expression of renal miRNAs and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service