Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9905

Sigma-Aldrich

Anti-Endothelin B Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ET-B Receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunocytochemistry: 4-11 μg/mL using human adrenal tissue
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-11 μg/mL using human adrenal tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... EDNRB(1910)
mouse ... Ednrb(13618)
rat ... Ednrb(50672)

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human endothelin B receptor, conjugated to KLH. The immunizing peptide has 79% homology with the mouse gene and 84% homology with the rat gene.

Application

Anti-Endothelin B Receptor antibody produced in rabbit is suitable for the following applications:
  • Immunocytochemistry at a concentration of 4-11μg/mL using human adrenal tissue
  • Immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 4-11μg/mL using human adrenal tissue
  • Western blotting
  • Immunofluorescence
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

EDNRB Endothelin receptor type B (ETB) is a protein encoded by the EDNRB gene in humans. It plays a pivotal role in oesophageal cancer and may act as a therapeutic target for this intractable malignancy. It has a novel role in oligodendroglioma proliferation and acts as a potential therapeutic alternative for oligodendrogliomas. Lower expression of ETB receptor responsiveness is associated with polycystic ovary syndrome (PCOS) and may reflect lower endothelial-mediated vasodilatation independent of generally lower vascular reactivity.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda J Leblanc et al.
Microcirculation (New York, N.Y. : 1994), 20(5), 365-376 (2012-12-04)
The risk for cardiovascular disease increases with advancing age; however, the chronological development of heart disease differs in males and females. The purpose of this study was to determine whether age-induced alterations in responses of coronary arterioles to the endogenous
J Milara et al.
British journal of pharmacology, 161(7), 1599-1615 (2010-08-12)
Pulmonary arteries from smokers and chronic obstructive pulmonary disease patients show abnormal endothelium-dependent vascular reactivity. We studied the effect of cigarette smoke extract (CSE) on endothelin receptor B (ET(B) ) expression in human pulmonary artery endothelial cells (HPAECs) and its
Suhas Vasaikar et al.
BMC cancer, 18(1), 154-154 (2018-02-08)
Glioblastoma (GBM) is the most common malignant brain tumor with median survival of 12-15 months. Owing to uncertainty in clinical outcome, additional prognostic marker(s) apart from existing markers are needed. Since overexpression of endothelin B receptor (ETBR) has been demonstrated in
T Tanaka et al.
British journal of cancer, 110(4), 1027-1033 (2013-12-21)
The endothelin axis has been shown to have a pivotal role in several human malignancies. The aim of this study was to clarify the clinical importance of endothelin receptor type B (ETBR) in human oesophageal squamous cell carcinoma (OSCC). We
Megan M Wenner et al.
The Journal of physiology, 589(Pt 19), 4671-4679 (2011-08-10)
Endothelin-1 is elevated in women with polycystic ovary syndrome (PCOS), and may play a role in the endothelial dysfunction associated with PCOS. Endothelin-1 binds two receptor subtypes, endothelin A (ET-A) and endothelin B (ET-B). We hypothesized that ET-A mediates vasoconstriction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service