Skip to Content
MilliporeSigma

Skip To

E8779

E-Toxate reagent from Limulus polyphemus

sufficient for 20 tests

Synonym(s):

Endotoxin Reagent, LAL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.26
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

dry concentrate

usage

sufficient for 20 tests

detection sensitivity measuring range

0.05-0.1 EU/mL

storage temp.

−20°C

Application

The endotoxins from Salmonella typhimurium isolated from human waste samples were detectable at a low concentration of 0.5 μg/ml using E-TOXATE reagent.

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E9029E4274T1949
usage

sufficient for 20 tests

usage

-

usage

-

usage

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

-

storage temp.

-

storage temp.

-

detection sensitivity measuring range

0.05-0.1 EU/mL

detection sensitivity measuring range

-

detection sensitivity measuring range

-

detection sensitivity measuring range

-

form

dry concentrate

form

-

form

-

form

-


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. What is the difference between Product No. E8779 and E8904, E-Toxate™ reagent from Limulus polyphemus?

    1 answer
    1. Both products are E-Toxate™ reagents from Limulus polyphemus. Product No. E8779 is sufficient for 20 tests and E8904 is sufficient for 50 tests.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. Is there a standard that can be used with E-Toxate™ reagent from Limulus polyphemus, Product E8779?

    1 answer
    1. A standard that can be used is Product No. E8029 (Sigma  E-Toxate™ Endotoxin standard).

      Helpful?

  4. How should E-Toxate™ reagent from Limulus polyphemus, Product E8779, be used?

    1 answer
    1. This product is a kit component of Product No. ET0200 (20 assay) and ET0300 (50 assay).

      Helpful?

Reviews

No rating value

Active Filters