Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8406

Sigma-Aldrich

Anti-Parvalbumin Antibody

rabbit polyclonal

Synonym(s):

Anti-EDEM, Anti-ER degradation enhancer, mannosidase alpha-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-EDEM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 65 kDa (nonglycosylated human and mouse EDEM1)
antigen 75 kDa

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 0.5-1.0 μg using extract HEK-293T cells expressing recombinant mouse EDEM1 protein.
western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells expressing recombinant human EDEM1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EDEM1(9695)
mouse ... Pvalb(19293)

General description

ER degradation-enhancing alpha-mannosidase-like 1 (EDEM1) is an enzyme encoded by the EDEM1 gene in humans. In mammalian cells, EDEM1 is localized to the endoplasmic reticulum (ER), mainly as a soluble glycoprotein.
glycoslylated human and mouse EDEM1

Immunogen

synthetic peptide corresponding to amino acids 629-646 of human EDEM1, conjugated to KLH via an N-terminal cysteine residue. The corresponding sequence is identical in mouse EDEM1.

Application

Anti-EDEM1 antibody has been used in immunoblotting.
Anti-EDEM1 antibody produced in rabbit is suitable for the following applications:
  • Immunoprecipitation at a concentration of 0.5-1.0μg using extract HEK-293T cells expressing recombinant mouse EDEM1 protein.
  • Western blotting at a concentration of 0.5-1.0μg/mL using whole extracts of HEK-293T cells expressing recombinant human EDEM1.
  • Immunofluorescence Microscopy.
  • Immunoblotting
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ER degradation-enhancing alpha-mannosidase-like 1 (EDEM1), a putative mannose binding lectin, targets misfolded glycoproteins for degradation in an N-glycan dependent manner. It interacts with the C-terminus of calnexin and lacks mannosidase activity.
ER degradation-enhancing alpha-mannosidase-like 1 is an enzyme encoded by the EDEM1 gene in humans. It is a chaperone for rod opsin and its expression can be used to promote correct folding enhancing degradation of mutant proteins in the ER to combat protein-misfolding disease. The endogenous EDEM1 in cells that are not stressed by the expression of a transgenic misfolded protein reaches the cytosol and is degraded by basal autophagy. EDEM may maintain the retrotranslocation competence of NHK by inhibiting aggregation. It is also considered responsible for directing aberrant proteins for ERAD (ER-associated protein degradation).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EDEM1 recognition and delivery of misfolded proteins to the SEL1L-containing ERAD complex
Cormier J H, et al.
Molecular Cell, 34(5), 627-633 (2009)
Sujin Park et al.
Histochemistry and cell biology, 142(2), 153-169 (2014-03-26)
ER degradation-enhancing α-mannosidase-like 1 protein (EDEM1) is involved in the routing of misfolded glycoproteins for degradation in the cytoplasm. Previously, we reported that EDEM1 leaves the endoplasmic reticulum via non-COPII vesicles (Zuber et al. in Proc Natl Acad Sci USA
Redundant and antagonistic roles of XTP3B and OS9 in decoding glycan and non-glycan degrons in ER-associated degradation
van der Goot A T, et al.
Molecular Cell, 70, 516-530 (2018)
Fulvio Reggiori et al.
Cell host & microbe, 7(6), 500-508 (2010-06-15)
Coronaviruses (CoV), including SARS and mouse hepatitis virus (MHV), are enveloped RNA viruses that induce formation of double-membrane vesicles (DMVs) and target their replication and transcription complexes (RTCs) on the DMV-limiting membranes. The DMV biogenesis has been connected with the
Redundant and Antagonistic Roles of XTP3B and OS9 in Decoding Glycan and Non-glycan Degrons in ER-Associated Degradation
van der Goot AT, et al.
Molecular Cell, 70(3), 516-530 e6-516-530 e6 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service