Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E6507

Sigma-Aldrich

β-Estradiol 6-(O-carboxy­methyl)oxime: BSA fluorescein iso­thio­cyanate conjugate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

extent of labeling

3-5 mol FITC per mol BSA
5-10 mol steroid per mol BSA

solubility

0.1 M phosphate buffer: 0.98-1.02 mg/mL, clear, yellow to orange (pH: 7.5)

shipped in

ambient

storage temp.

2-8°C

SMILES string

S=C=Nc5c6c(ccc5O)C8(OC(=O)c9c8cccc9)c7c(cc(cc7)O)O6.N(OCC(=O)O)C1=C[C@H]2[C@H]3[C@@]([C@H](CC3)O)(CC[C@@H]2c4c1cc(cc4)O)C

InChI key

DZYKSUALQNKEPM-TUXUTZNTSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yu Pan et al.
Hormones & cancer, 11(2), 87-96 (2020-02-11)
Proline-, glutamic acid-, leucine-rich protein 1 (PELP1) is a novel estrogen receptor (ER) coregulator, demonstrated distinctive characters from other ERα coregulators, and has been suggested to be involved in metastasis of several cancers. In ERα-positive breast cancer, PELP1 overexpression enhanced
Synthesis of fluorescein labelled steroid hormone-albumin conjugates for the fluorescent histochemical detection of hormone receptors.
E Gaetjens et al.
Journal of steroid biochemistry, 13(8), 1001-1003 (1980-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service