Skip to Content
MilliporeSigma

Skip To

E5411

Enniatin B

From Gnomonia errabunda, ≥95% (HPLC), antibiotic

Synonym(s):

N-Methylcyclo(L-Val-D-Hmb-N-methyl-L-Val-D-Hmb-N-methyl-L-Val-D-Hmb-);(3S,6R,9S,12R,15S,18R)-3,6,9,12,15,18-Hexaisopropyl-4,10,16-trimethyl-1,7,13-trioxa-4,10,16-triazacyclooctadecane-2,5,8,11,14,17-hexone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C33H57N3O9
CAS Number:
Molecular Weight:
639.82
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Enniatin B, from Gnomonia errabunda, ≥95% (HPLC)

biological source

Gnomonia errabunda

Quality Segment

assay

≥95% (HPLC)

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

storage temp.

−20°C

SMILES string

N1([C@H](C(=O)O[C@@H](C(=O)N([C@H](C(=O)O[C@@H](C(=O)N([C@H](C(=O)O[C@@H](C1=O)C(C)C)C(C)C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C

InChI

1S/C33H57N3O9/c1-16(2)22-31(40)43-26(20(9)10)29(38)35(14)24(18(5)6)33(42)45-27(21(11)12)30(39)36(15)23(17(3)4)32(41)44-25(19(7)8)28(37)34(22)13/h16-27H,1-15H3/t22-,23-,24-,25+,26+,27+/m0/s1

InChI key

MIZMDSVSLSIMSC-VYLWARHZSA-N

Biochem/physiol Actions

Enniatins are a group of cyclohexadepsipeptide mycotoxins produced by Gnomonia errabuda and several Fusaria species, with phytotoxic, antibiotic, and insecticidal activities. Enniatins function as ionophors by their incorporation into the cellular membrane to form dimeric structures that transport monovalent ions across the membrane (especially the mitochondrial membranes) affecting oxidative phosphorylation uncoupling. It has been demonstrated that enniatins have a cytotoxic effect on human cancer cells. Furthermore, incubation of H4IIE hepatoma cells with enniatins strongly diminished phosphorylation of the ERK (p44/p42). Enniatins B and B1 inhibit the multi-drug resistance transporter Pdr5p from Saccharomyces cerevisiae, indicating their beneficial potential in cases of drug resistant patients.
Enniatins are cyclohexadepsipeptide mycotoxins that have phytotoxic, antibiotic, and insecticidal activities and function as ionophors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E5286E9661E5161
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters