Skip to Content

E4260

β-Estradiol-d2

98 atom % D

Synonym(s):

1,3,5(10)-Estratriene-2,4-d2-3,17β-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H22D2O2
CAS Number:
Molecular Weight:
274.39
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352115
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

98 atom % D

Quality Segment

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4c([2H])c(O)c([2H])cc24

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3D,10D

InChI key

VOXZDWNPVJITMN-JXCLBMSPSA-N

General description

β-Estradiol-d2 98 atom % D is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- Chlorination reaction mechanism studies
- reaction mechanism investigations
- Analytical method development
- Pharmaceutical formulation development

Features and Benefits

Features
- Ideal for mass spectrometry applications
- Enhances the sensitivity of analytical techniques
- Supports high-throughput screening methods
- Safe and effective for laboratory use

Benefits
- Increases the sensitivity of mass spectrometry
- Provides valuable insights for pharmacokinetics studies
- Enhances the understanding of protein interactions
- Supports innovative research in structural biology


Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library