Skip to Content
MilliporeSigma

E2750

(+)-Pseudoephedrine hydrochloride

≥98%

Synonym(s):

(+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HCl
CAS Number:
Molecular Weight:
201.69
UNSPSC Code:
12352210
NACRES:
NA.77
PubChem Substance ID:
EC Number:
206-462-1
Beilstein/REAXYS Number:
3915112
MDL number:
Assay:
≥98%
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

185-188 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

Cl.CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10+;/m0./s1

InChI key

BALXUFOVQVENIU-KXNXZCPBSA-N

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library