Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E2656

Sigma-Aldrich

(−)-Epothilone B

from Sorangium cellulosum, ≥98% (HPLC)

Synonym(s):

EPO906, EpoB, Patupilone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H41NO6S
CAS Number:
Molecular Weight:
507.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Sorangium cellulosum

Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

storage condition

desiccated
protect from light

color

white to off-white

storage temp.

−20°C

SMILES string

[H][C@]1(C[C@]2([H])O[C@]2(C)CCC[C@H](C)[C@H](O)[C@@H](C)C(=O)C(C)(C)[C@@H](O)CC(=O)O1)\C(C)=C\c3csc(C)n3

InChI

1S/C27H41NO6S/c1-15-9-8-10-27(7)22(34-27)12-20(16(2)11-19-14-35-18(4)28-19)33-23(30)13-21(29)26(5,6)25(32)17(3)24(15)31/h11,14-15,17,20-22,24,29,31H,8-10,12-13H2,1-7H3/b16-11+/t15-,17+,20-,21-,22-,24-,27+/m0/s1

InChI key

QXRSDHAAWVKZLJ-PVYNADRNSA-N

Related Categories

General description

Epothilones has antimitotic properties. It prevents microtubule depolymerization and competitively blocks paclitaxel binding to microtubules. Epothilone B is used to treat metastatic breast cancer (MBC).

Biochem/physiol Actions

(-)-Epothilone B is a microtubule (MT) stabilizing drug and natural macrolide antitumor from myxobacteria Sorangium cellulosum. EpoB has similar biological properties to EpoA. However, EpoB is 10-fold more potent than EpoA against P-glycoprotein-expressing multidrug resistant (MDR) cells (IC50 = 2 nM for MDR CCRF-CEM/VBL100 cells). (-)-Epothilone B is similar to paclitaxel in binding displacement, and a substitution for paclitaxel in dependent cell growth. EpoB causes cell cycle arrest (IC50 = 3.5 nM).
EpoB is a microtubule stabilizing drug and natural macrolide antitumor.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase II clinical trial of ixabepilone (BMS-247550), an epothilone B analog, as first-line therapy in patients with metastatic breast cancer previously treated with anthracycline chemotherapy
Roche H, et al.
Journal of Clinical Oncology, 25(23), 3415-3420 (2007)
BMS-247550: a novel epothilone analog with a mode of action similar to paclitaxel but possessing superior antitumor efficacy
Lee FYF, et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 7(5), 1429-1437 (2001)
Christoph Grohmann et al.
Cell death & disease, 12(3), 268-268 (2021-03-14)
Targeting cell division by chemotherapy is a highly effective strategy to treat a wide range of cancers. However, there are limitations of many standard-of-care chemotherapies: undesirable drug toxicity, side-effects, resistance and high cost. New small molecules which kill a wide
Colette J Shen et al.
BMC cancer, 14, 532-532 (2014-07-24)
Ionizing radiation (IR) is a mainstay of cancer therapy, but irradiation can at times also lead to stress responses, which counteract IR-induced cytotoxicity. IR also triggers cellular secretion of vascular endothelial growth factor, transforming growth factor β and matrix metalloproteinases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service