Skip to Content
MilliporeSigma

E0518

C1 Esterase Inhibitor

aqueous solution, ≥95% (SDS-PAGE)

Synonym(s):

Complement C1 esterase inhibitor, Esterase inhibitor C-1 from human plasma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

C1 Esterase Inhibitor from human plasma, aqueous solution, ≥95% (SDS-PAGE)

biological source

human plasma

Quality Segment

assay

≥95% (SDS-PAGE)

form

aqueous solution

mol wt

~105 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SERPING1(710)

Biochem/physiol Actions

Esterase inhibitor C1 is a multifunctional regulator of all major kinin-generating protein cascade systems. It has been a therapeutic tool in the treatment of hereditary angioedema and complement-mediated inflammatory tissue damage such as capillary leak syndrome, septic shock, multiple organ failure, and hyperacute graft rejection.

Physical form

Solution in 20 mM potassium phosphate, pH 7.0, containing 250 mM KCl

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
E0518-1MG04061826682500