Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DUO82008

Sigma-Aldrich

Duolink Antibody Diluent (1X)

Synonym(s):

Antibody diluent, Dilution buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

form

liquid

Quality Level

shipped in

wet ice

storage temp.

2-8°C

General description

Duolink antibody Diluent for Duolink PLA assay

Application

Duolink Antibody Diluent (1X) has been used in situ proximity ligation assay (PLA).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service