Skip to Content
MilliporeSigma

DMPS5X

Dimethylpolysiloxane

viscosity 50 cSt (25 °C)(lit.)

Synonym(s):

Polydimethylsiloxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

mol wt

~3,780

viscosity

50 cSt(25 °C)(lit.)

SMILES string

[Si](=O)(C)C

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Application

Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
Dimethylpolysiloxane has been used:
  • in culturing mouse embryos
  • in preparation of oil-based nano-adjuvant candidates
  • to reduce the brain movement in the post-cat craniotomy process

Biochem/physiol Actions

Dimethylpolysiloxane is a silicon-based organic polymer, inert and non-toxic. It is applicable in the pharmaceutical, food, and cosmetic industry. Dimethylpolysiloxane is excreted unchanged unaltered in the faeces and is non-mutagenic.
Dimethylpolysiloxane is used in protein chromatography and affininty chromatography. Dimethylpolysiloxane was used to determine that postprandial inflammatory response after ingestion of heated oils in obese persons is reduced by the presence of phenol compounds.


Still not finding the right product?

Explore all of our products under Dimethylpolysiloxane


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

609.8 °F - closed cup

flash_point_c

321 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library