Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D8013

Sigma-Aldrich

P1,P5-Di(adenosine-5′) pentaphosphate ammonium salt

≥95% (HPLC), powder

Synonym(s):

A(5′)P5(5′)A, Diadenosine pentaphosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N10O22P5 · xNH3 · yH2O
CAS Number:
Molecular Weight:
916.37 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](COP(OP(O)(OP(O)(OP(O)(OP(O)(OC[C@@H]2[C@@H](O)[C@@H](O)[C@H](N(C=N3)C4=C3C(N)=NC=N4)O2)=O)=O)=O)=O)(O)=O)O[C@@H](N(C=N5)C6=C5C(N)=NC=N6)[C@@H]1O

InChI

1S/C10H14N5O8P3/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(21-7)3-20-25(16,17)23-26(18,19)22-24/h1-2,4-7H,3,24H2,(H,16,17)(H,18,19)(H2,11,12,13)/t6-,7+/m0/s1

InChI key

LWOSDYWLTWLFID-NKWVEPMBSA-N

Related Categories

Biochem/physiol Actions

A diadenosine polyphosphate stored in secretory granules of thrombocytes, chromaffin and neuronal cells.
A diadenosine polyphosphate stored in secretory granules of thrombocytes, chromaffin and neuronal cells. After release into the extracellular space, it affects a variety of biological activities in a wide range of target tissues. In the nervous system it acts through various purinergic receptors. It also activates 5′-nucleotidase and inhibits adenosine kinase activity in vitro. Ap5A is metabolized by soluble enzymes in the blood plasma and by membrane-bound ectoenzymes of a number of cell types including endothelial and smooth muscle cells. In heart, it increases contractile force and action potential duration, and shows an anti-β-adrenergic effect.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Brewer et al.
Journal of neurochemistry, 74(5), 1968-1978 (2000-05-09)
The loss of ATP, which is needed for ionic homeostasis, is an early event in the neurotoxicity of glutamate and beta-amyloid (A(beta)). We hypothesize that cells supplemented with the precursor creatine make more phosphocreatine (PCr) and create larger energy reserves
Yapeng Ji et al.
Nature communications, 8, 15308-15308 (2017-05-19)
Targeting the specific metabolic phenotypes of colorectal cancer stem cells (CRCSCs) is an innovative therapeutic strategy for colorectal cancer (CRC) patients with poor prognosis and relapse. However, the context-dependent metabolic traits of CRCSCs remain poorly elucidated. Here we report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service