Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4908

Sigma-Aldrich

Anti-Dog Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, dog

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

canine

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

Strong reactivity with normal dog serum has been determined. This antiserum has not been assayed for interspecies cross reactivity.

Application

Anti-Dog Serum antibody produced in rabbit has been used in immunochemistry to identify the feeding source in the promesenteron of male and female insects.

Biochem/physiol Actions

Adult dog serum can be used as a substitute for colostrum in the neonatal dog.

Physical form

Rabbit Anti-Dog Serum is provided as a liquid containing 0.1% sodium azide as preservative.

Storage and Stability

For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of adult dog serum as a substitute for colostrum in the neonatal dog.
Poffenbarger EM, et al.
American Journal of Veterinary Research, 52(8), 1221-1224 (1991)
Host influence on the nutritional and reproductive status of Triatoma infestans (Klug)(Hemiptera: Reduviidae) peridomiciliary populations
Soria C, et al.
Revista de la Sociedad Entomologica Argentina, 78(2) (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service